Anti-SIRT3 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
S1013-88F4FITC
USBIS1013-88F4FITCEA 884 EUR
USBIS1013-88F4FITC
Anti-SIRT3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: SIRT3 (sirtuin 3)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR