Anti-SKI Mouse Monoclonal Antibody [clone: 6D763]

Lieferant: US Biological
S1013-89M
USBIS1013-89MEA 611 EUR
USBIS1013-89M
Anti-SKI Mouse Monoclonal Antibody [clone: 6D763]
Antikörper
Recommended dilution: Immunochemistry: 1-10 ?g/ml; Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: SKI (v-ski avian sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone: 6D763
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rat, Xenopus laevis
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR