Anti-SKI Rabbit Polyclonal Antibody

Lieferant: US Biological
S1013-89V
USBIS1013-89VEA 701 EUR
USBIS1013-89V
Anti-SKI Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 2-2.5 ?g/ml

Type: Primary
Antigen: SKI (v-ski avian sarcoma viral oncogene homolog)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR