Anti-Ski-like Protein Rabbit Polyclonal Antibody

Lieferant: US Biological
S1013-89W
USBIS1013-89WEA 701 EUR
USBIS1013-89W
Anti-Ski-like Protein Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-3?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: Ski-like Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR