Anti-SKI Mouse Monoclonal Antibody

Lieferant: US Biological
S1013-89Z3
USBIS1013-89Z3EA 590 EUR
USBIS1013-89Z3
Anti-SKI Mouse Monoclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: SKI (v-ski avian sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR