Anti-Smoothened Rabbit Polyclonal Antibody
Lieferant: US Biological
S1014-63E
USBIS1014-63EEA 513 EUR
USBIS1014-63E
Anti-Smoothened Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.1-1 ?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: Smoothened
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse