Anti-Sodium-dependent Vitamin C Transporter 1 Rabbit Polyclonal Antibody

Lieferant: US Biological
S5327-40-100
USBIS5327-40-100EA 742 EUR
USBIS5327-40-100
Anti-Sodium-dependent Vitamin C Transporter 1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: Sodium-dependent Vitamin C Transporter 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR