Anti-APCS Rabbit Polyclonal Antibody

Lieferant: US Biological
S5451-20
USBIS5451-20EA 649 EUR
USBIS5451-20
Anti-APCS Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: APCS (amyloid P component, serum)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR