Anti-SPRY4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
S5620-25B-APC
USBIS5620-25B-APCEA 884 EUR
USBIS5620-25B-APC
Anti-SPRY4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000 Dot Blot: 1:500
Do not freeze!

Type: Primary
Antigen: SPRY4 (sprouty homolog 4 (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR