Anti-SPRY4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
S5620-25B-PE
USBIS5620-25B-PEEA 884 EUR
USBIS5620-25B-PE
Anti-SPRY4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000 Dot Blot: 1:500
Do not freeze!
Type: Primary
Antigen: SPRY4 (sprouty homolog 4 (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human