Anti-SRC Mouse Monoclonal Antibody [clone: 10i71]

Lieferant: US Biological
S6500-10B
USBIS6500-10BEA 649 EUR
USBIS6500-10B
Anti-SRC Mouse Monoclonal Antibody [clone: 10i71]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100; Immunoprecipitation: 2?g/test

Type: Primary
Antigen: SRC (v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog)
Clonality: Monoclonal
Clone: 10i71
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR