Anti-Src-like Adaptor Rabbit Polyclonal Antibody

Lieferant: US Biological
S6504-85B
USBIS6504-85BEA 466 EUR
USBIS6504-85B
Anti-Src-like Adaptor Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: Src-like Adaptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR