Anti-SSA1/Ro-52 Rabbit Polyclonal Antibody

Lieferant: US Biological
S6600-65-100
USBIS6600-65-100EA 418 EUR
USBIS6600-65-100
Anti-SSA1/Ro-52 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:125000

Type: Primary
Antigen: SSA1/Ro-52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rodent
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR