Anti-Stearoyl CoA Desaturase 1 Rabbit Monoclonal Antibody [clone: 9j153]

Lieferant: US Biological
S7900-05B
USBIS7900-05BEA 614 EUR
USBIS7900-05B
Anti-Stearoyl CoA Desaturase 1 Rabbit Monoclonal Antibody [clone: 9j153]
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50; Immunochemistry: 1:200; Immunofluorescence: 1:100

Type: Primary
Antigen: Stearoyl CoA Desaturase 1
Clonality: Monoclonal
Clone: 9j153
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR