Anti-Stearoyl CoA Desaturase 1 Rabbit Polyclonal Antibody

Lieferant: US Biological
S7900-06
USBIS7900-06EA 531 EUR
USBIS7900-06
Anti-Stearoyl CoA Desaturase 1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:2000-1:5000; Western Blot: 1:2000-1:5000

Type: Primary
Antigen: Stearoyl CoA Desaturase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR