Anti-STAT 1 Mouse Monoclonal Antibody [clone: 5J111]

Lieferant: US Biological
S7969-05G
USBIS7969-05GEA 809 EUR
USBIS7969-05G
Anti-STAT 1 Mouse Monoclonal Antibody [clone: 5J111]
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: STAT 1
Clonality: Monoclonal
Clone: 5J111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR