Anti-Stat 2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
S7969-83A-BIOT
USBIS7969-83A-BIOTEA 884 EUR
USBIS7969-83A-BIOT
Anti-Stat 2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Stat 2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR