Anti-STAT3 Mouse Monoclonal Antibody [clone: 3G275]

Lieferant: US Biological
S7971-03X
USBIS7971-03XEA 485 EUR
USBIS7971-03X
Anti-STAT3 Mouse Monoclonal Antibody [clone: 3G275]
Antikörper
Recommended dilution: ELISA: 0.1-1 ?g/ml; Western Blot: 1-2?g/ml

Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: Monoclonal
Clone: 3G275
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR