Anti-STAT3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
S7971-20B-APC
USBIS7971-20B-APCEA 884 EUR
USBIS7971-20B-APC
Anti-STAT3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human