Anti-STAT3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: US Biological
S7971-20C-AP
USBIS7971-20C-APEA 884 EUR
USBIS7971-20C-AP
Anti-STAT3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human