Anti-STAT3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
S7971-20C-APC
USBIS7971-20C-APCEA 884 EUR
USBIS7971-20C-APC
Anti-STAT3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR