Anti-Stat 5b Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
S7972-07S-FITC
USBIS7972-07S-FITCEA 884 EUR
USBIS7972-07S-FITC
Anti-Stat 5b Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: Stat 5b
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR