Anti-Stat 5a Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
S7972-10B-APC
USBIS7972-10B-APCEA 884 EUR
USBIS7972-10B-APC
Anti-Stat 5a Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100; Dot Blot: 1:500
Do not freeze!

Type: Primary
Antigen: Stat 5a
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR