Anti-STAT 5a Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 7H57]

Lieferant: US Biological
S7972-10J
USBIS7972-10JEA 654 EUR
USBIS7972-10J
Anti-STAT 5a Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 7H57]
Antikörper
Type: Primary
Antigen: STAT 5a
Clonality: Monoclonal
Clone: 7H57
Conjugation: Carboxyfluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR