Anti-STAT 5a Rabbit Polyclonal Antibody

Lieferant: US Biological
S7972-11F
USBIS7972-11FEA 425 EUR
USBIS7972-11F
Anti-STAT 5a Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: STAT 5a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR