Anti-Serine/threonine kinase 11, STK11 Rabbit Polyclonal Antibody

Lieferant: US Biological
S7973-58W4
USBIS7973-58W4EA 450 EUR
USBIS7973-58W4
Anti-Serine/threonine kinase 11, STK11 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: Serine/threonine kinase 11, STK11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR