Anti-STK11 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
S7973-58Y7FITC
USBIS7973-58Y7FITCEA 884 EUR
USBIS7973-58Y7FITC
Anti-STK11 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: STK11 (serine/threonine kinase 11)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR