Anti-Streptococcus Group A Rabbit Polyclonal Antibody

Lieferant: US Biological
S7974-02C
USBIS7974-02CEA 383 EUR
USBIS7974-02C
Anti-Streptococcus Group A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Streptococcus Group A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR