Anti-Streptococcus Group A Mouse Monoclonal Antibody [clone: 10F468]

Lieferant: US Biological
S7974-02G
USBIS7974-02GEA 1190 EUR
USBIS7974-02G
Anti-Streptococcus Group A Mouse Monoclonal Antibody [clone: 10F468]
Antikörper
Type: Primary
Antigen: Streptococcus Group A
Clonality: Monoclonal
Clone: 10F468
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR