Anti-Streptococcus Group A Mouse Monoclonal Antibody [clone: 9L667]
Lieferant: US Biological
S7974-50A
USBIS7974-50AEA 1450 EUR
USBIS7974-50A
Anti-Streptococcus Group A Mouse Monoclonal Antibody [clone: 9L667]
Antikörper
Type: Primary
Antigen: Streptococcus Group A
Clonality: Monoclonal
Clone: 9L667
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen