Anti-Substance P Mouse Monoclonal Antibody [clone: SP-DE4-21]

Lieferant: US Biological
S8000-01B-100
USBIS8000-01B-100EA 643 EUR
USBIS8000-01B-100
Anti-Substance P Mouse Monoclonal Antibody [clone: SP-DE4-21]
Antikörper
Recommended dilution: ELISA: 1:10000; Immunochemistry: 1:300-1:1000

Type: Primary
Antigen: Substance P
Clonality: Monoclonal
Clone: SP-DE4-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Guinea pig, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR