Anti-UBA2 Rabbit Polyclonal Antibody

Lieferant: US Biological
S8025-90H
USBIS8025-90HEA 466 EUR
USBIS8025-90H
Anti-UBA2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: UBA2 (ubiquitin-like modifier activating enzyme 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR