Anti-UBA2 Rabbit Polyclonal Antibody

Lieferant: US Biological
S8025-90K
USBIS8025-90KEA 633 EUR
USBIS8025-90K
Anti-UBA2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.25-0.5 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: UBA2 (ubiquitin-like modifier activating enzyme 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR