Anti-UBA2 Goat Polyclonal Antibody

Lieferant: US Biological
S8025-90L
USBIS8025-90LEA 601 EUR
USBIS8025-90L
Anti-UBA2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000

Type: Primary
Antigen: UBA2 (ubiquitin-like modifier activating enzyme 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR