Anti-SUMO, Pan Rabbit Polyclonal Antibody

Lieferant: US Biological
S8026-01A
USBIS8026-01AEA 667 EUR
USBIS8026-01A
Anti-SUMO, Pan Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: SUMO, Pan
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR