Anti-SUMO, Pan Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
S8026-01B-BIOT
USBIS8026-01B-BIOTEA 884 EUR
USBIS8026-01B-BIOT
Anti-SUMO, Pan Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: SUMO, Pan
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR