Anti-Sulfadoxine Sheep Polyclonal Antibody

Lieferant: US Biological
S8029
USBIS8029EA 763 EUR
USBIS8029
Anti-Sulfadoxine Sheep Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 5 ?g/ml

Type: Primary
Antigen: Sulfadoxine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR