Anti-SYN1 Rabbit Monoclonal Antibody [clone: 10g91]

Lieferant: US Biological
S9104-01J
USBIS9104-01JEA 647 EUR
USBIS9104-01J
Anti-SYN1 Rabbit Monoclonal Antibody [clone: 10g91]
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:800; Immunofluorescence: 1:200; Immunoprecipitation: 1:50

Type: Primary
Antigen: SYN1 (synapsin I)
Clonality: Monoclonal
Clone: 10g91
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR