Anti-SYT4 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
S9109-40B
USBIS9109-40BEA 836 EUR
USBIS9109-40B
Anti-SYT4 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: SYT4 (synaptotagmin IV)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR