Anti-TAFA2 Sheep Polyclonal Antibody

Lieferant: US Biological
T0600-52
USBIT0600-52EA 856 EUR
USBIT0600-52
Anti-TAFA2 Sheep Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 5-15 ?g/ml

Type: Primary
Antigen: TAFA2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR