Anti-Tamm-Horsfall Urinary Glycoprotein Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10.32]

Lieferant: US Biological
T0850-03
USBIT0850-03EA 701 EUR
USBIT0850-03
Anti-Tamm-Horsfall Urinary Glycoprotein Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10.32]
Antikörper
Recommended dilution: Immunochemistry: 1:50

Type: Primary
Antigen: Tamm-Horsfall Urinary Glycoprotein
Clonality: monoclonal
Clone: 10.32
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR