Anti-TNKS2 Goat Polyclonal Antibody

Lieferant: US Biological
T0900-09D
USBIT0900-09DEA 640 EUR
USBIT0900-09D
Anti-TNKS2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1. 5 ?g/ml

Type: Primary
Antigen: TNKS2 (tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR