Anti-TBX4 Rabbit Polyclonal Antibody

Lieferant: US Biological
T1848-26T
USBIT1848-26TEA 568 EUR
USBIT1848-26T
Anti-TBX4 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TBX4 (T-box 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR