Anti-T-complex Protein 1 eta Rat Monoclonal Antibody [clone: 10B491]

Lieferant: US Biological
T2035-35
USBIT2035-35EA 641 EUR
USBIT2035-35
Anti-T-complex Protein 1 eta Rat Monoclonal Antibody [clone: 10B491]
Antikörper
Recommended dilution: Immunochemistry: 1:100; Western Blot: 1:500

Type: Primary
Antigen: T-complex Protein 1 eta
Clonality: monoclonal
Clone: 10B491
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR