Anti-TCR, alpha, beta Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: H57-597]
Lieferant: US Biological
T2040-02N3
USBIT2040-02N3EA 339 EUR
USBIT2040-02N3
Anti-TCR, alpha, beta Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: H57-597]
Antikörper
Do not freeze!
Type: Primary
Antigen: TCR, alpha, beta
Clonality: monoclonal
Clone: H57-597
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse