Anti-TCR, alpha, beta Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: H57-597]

Lieferant: US Biological
T2040-02N5
USBIT2040-02N5EA 405 EUR
USBIT2040-02N5
Anti-TCR, alpha, beta Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: H57-597]
Antikörper
Type: Primary
Antigen: TCR, alpha, beta
Clonality: monoclonal
Clone: H57-597
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR