Anti-TCR, alpha, beta, V beta 2 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K83]

Lieferant: US Biological
T2040-02W1
USBIT2040-02W1EA 582 EUR
USBIT2040-02W1
Anti-TCR, alpha, beta, V beta 2 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K83]
Antikörper
Type: Primary
Antigen: TCR, alpha, beta, V beta 2
Clonality: monoclonal
Clone: 5K83
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR