Anti-TECPR1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: US Biological
T2275-80N-AP
USBIT2275-80N-APEA 884 EUR
USBIT2275-80N-AP
Anti-TECPR1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: TECPR1 (tectonin beta-propeller repeat containing 1)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human