Anti-TET2 Goat Polyclonal Antibody

Lieferant: US Biological
T2958-60A
USBIT2958-60AEA 601 EUR
USBIT2958-60A
Anti-TET2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000

Type: Primary
Antigen: TET2 (tet methylcytosine dioxygenase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR