Anti-Theophylline Mouse Monoclonal Antibody [clone: 9L702]

Lieferant: US Biological
T4500-03B
USBIT4500-03BEA 540 EUR
USBIT4500-03B
Anti-Theophylline Mouse Monoclonal Antibody [clone: 9L702]
Antikörper
Type: Primary
Antigen: Theophylline
Clonality: monoclonal
Clone: 9L702
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR